[5-(Trifluoromethyl)-1,2,4-oxadiazol-3-yl]methanol

Supplier: Matrix Scientific

077812-1G
102667-876EA 649.23 USD
102667-876
[5-(Trifluoromethyl)-1,2,4-oxadiazol-3-yl]methanol
[5-(Trifluoromethyl)-1,2,4-oxadiazol-3-yl]methanol

Hazard: Irritant

Formula: C₄H₃F₃N₂O₂
MDL Number: MFCD11847842
CAS Number: 1260674-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR