Methyl (2Z)-3-(3-bromo-1H-1,2,4-triazol-5-yl)acrylate

Supplier: Matrix Scientific

077786-500MG
102667-746EA 516.92 USD
102667-746
Methyl (2Z)-3-(3-bromo-1H-1,2,4-triazol-5-yl)acrylate
Methyl (2Z)-3-(3-bromo-1H-1,2,4-triazol-5-yl)acrylate

Hazard: Irritant

Formula: C₆H₆BrN₃O₂
MDL Number: MFCD22056546
CAS Number: 1365988-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR