5-(Ethylsulfonyl)-1,3-benzoxazol-2-amine

Supplier: Matrix Scientific

077758-500MG
102667-596EA 516.92 USD
102667-596
5-(Ethylsulfonyl)-1,3-benzoxazol-2-amine
5-(Ethylsulfonyl)-1,3-benzoxazol-2-amine

Hazard: Irritant

Formula: C₉H₁₀N₂O₃S
MDL Number: MFCD06655668
CAS Number: 852851-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR