1-Ethyl-3,3,7-trimethyl-5-nitro-1,3-dihydro-2H-indol-2-one

Supplier: Matrix Scientific

077749-500MG
102667-560EA 258.46 USD
102667-560
1-Ethyl-3,3,7-trimethyl-5-nitro-1,3-dihydro-2H-indol-2-one
1-Ethyl-3,3,7-trimethyl-5-nitro-1,3-dihydro-2H-indol-2-one

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₃
MDL Number: MFCD17265872
CAS Number: 1248559-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR