4-Isopropoxy-1-benzofuran-6-carboxylic acid

Supplier: Matrix Scientific

077694-1G
102667-500EA 646.15 USD
102667-500
4-Isopropoxy-1-benzofuran-6-carboxylic acid
4-Isopropoxy-1-benzofuran-6-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₂O₄
MDL Number: MFCD14704813
CAS Number: 1239773-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR