7-Methoxy-4,5-dihydro-1H-benzo-[g]indazole-3-carboxylic acid

Supplier: Matrix Scientific

077686-500MG
102667-456EA 387.69 USD
102667-456
7-Methoxy-4,5-dihydro-1H-benzo-[g]indazole-3-carboxylic acid
7-Methoxy-4,5-dihydro-1H-benzo-[g]indazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₃
MDL Number: MFCD03834491
CAS Number: 696645-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR