4-(Chloromethyl)-1,5-dimethyl-1H-pyrazole hydrochloride
Supplier: Matrix Scientific
077641-1G
077641-5G
077641-500MG
102667-270EA
1700
USD
102667-270
102667-266
102667-268
4-(Chloromethyl)-1,5-dimethyl-1H-pyrazole hydrochloride
4-(Chloromethyl)-1,5-dimethyl-1H-pyrazole hydrochloride
Hazard: Irritant
Formula:
C₆H₁₀Cl₂N₂ |
MDL Number:
MFCD20441439 CAS Number: 1390655-08-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...