7-Hydroxybicyclo[3.3.1]nonane-3-carbohydrazide hydrochloride

Supplier: Matrix Scientific

077619-5G 077619-500MG 077619-1G
102667-144EA 646.15 USD
102667-144 102667-146 102667-148
7-Hydroxybicyclo[3.3.1]nonane-3-carbohydrazide hydrochloride
7-Hydroxybicyclo[3.3.1]nonane-3-carbohydrazide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₉ClN₂O₂
MDL Number: MFCD20441451
CAS Number: 1401425-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR