1-(2-Chloro-5-fluoropyrimidin-4-yl)piperidin-4-amine dihydrochloride
Supplier: Matrix Scientific
077595-500MG
102667-032EA
516.92
USD
102667-032
1-(2-Chloro-5-fluoropyrimidin-4-yl)piperidin-4-amine dihydrochloride
1-(2-Chloro-5-fluoropyrimidin-4-yl)piperidin-4-amine dihydrochloride
Hazard: Irritant
Formula:
C₉H₁₄Cl₃FN₄ |
MDL Number:
MFCD22421959 CAS Number: 1401425-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...