3-(2-Furyl)-1H-1,2,4-triazole

Supplier: Matrix Scientific

077585-5G 077585-500MG 077585-1G
102667-000EA 484.62 USD
102667-000 102667-002 102666-998
3-(2-Furyl)-1H-1,2,4-triazole
3-(2-Furyl)-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₆H₅N₃O
MDL Number: MFCD18784752
CAS Number: 23195-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR