5-(2-Methylphenyl)piperidin-2-one
Supplier: Matrix Scientific
077455-1G
077455-5G
077455-500MG
102666-342EA
461.54
USD
102666-342
102666-340
102666-344
5-(2-Methylphenyl)piperidin-2-one
5-(2-Methylphenyl)piperidin-2-one
Hazard: Irritant
Formula:
C₁₂H₁₅NO |
MDL Number:
MFCD02926448 CAS Number: 41216-04-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...