3,5-Dimethyl-1H-pyrazole-4-carbohydrazide
Supplier: Matrix Scientific
077442-1G
077442-500MG
077442-5G
102666-276EA
484.62
USD
102666-276
102666-274
102666-278
3,5-Dimethyl-1H-pyrazole-4-carbohydrazide
3,5-Dimethyl-1H-pyrazole-4-carbohydrazide
Hazard: Irritant
Formula:
C₆H₁₀N₄O |
MDL Number:
MFCD07437052 CAS Number: 98484-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...