3-(1-Hydrazinoethyl)pyridine hydrochloride
Supplier: Matrix Scientific
077436-1G
077436-5G
077436-500MG
102666-242EA
2430.77
USD
102666-242
102666-240
102666-238
3-(1-Hydrazinoethyl)pyridine hydrochloride
3-(1-Hydrazinoethyl)pyridine hydrochloride
Hazard: Irritant
Formula:
C₇H₁₂ClN₃ |
MDL Number:
MFCD22421958 CAS Number: 1401426-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...