3-[4-(Ethoxycarbonyl)-1,3,5-trimethyl-1H-pyrrol-2-yl]propanoic acid

Supplier: Matrix Scientific

077428-1G 077428-5G 077428-500MG
102666-194EA 484.62 USD
102666-194 102666-192 102666-196
3-[4-(Ethoxycarbonyl)-1,3,5-trimethyl-1H-pyrrol-2-yl]propanoic acid
3-[4-(Ethoxycarbonyl)-1,3,5-trimethyl-1H-pyrrol-2-yl]propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₉NO₄
MDL Number: MFCD16992086
CAS Number: 1283390-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR