2-[(4-Amino-3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoic acid
Supplier: Matrix Scientific
077415-1G
077415-500MG
102666-122EA
484.62
USD
102666-122
102666-120
2-[(4-Amino-3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoic acid
2-[(4-Amino-3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoic acid
Hazard: Irritant
Formula:
C₁₃H₁₅N₃O₂ |
MDL Number:
MFCD20441350 CAS Number: 1338495-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...