3-[(2-Bromophenoxy)methyl]-4-methoxybenzoic acid
Supplier: Matrix Scientific
077410-1G
077410-500MG
102666-096EA
258.46
USD
102666-096
102666-098
3-[(2-Bromophenoxy)methyl]-4-methoxybenzoic acid
3-[(2-Bromophenoxy)methyl]-4-methoxybenzoic acid
Hazard: Irritant
Formula:
C₁₅H₁₃BrO₄ |
MDL Number:
MFCD03421969 CAS Number: 438218-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...