5-[(4-Chloro-2-methylphenoxy)methyl]-2-furoic acid
Supplier: Matrix Scientific
077406-5G
077406-1G
077406-500MG
102666-074EA
484.62
USD
102666-074
102666-072
102666-076
5-[(4-Chloro-2-methylphenoxy)methyl]-2-furoic acid
5-[(4-Chloro-2-methylphenoxy)methyl]-2-furoic acid
Hazard: Irritant
Formula:
C₁₃H₁₁ClO₄ |
MDL Number:
MFCD03421879 CAS Number: 832739-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...