[(5-Isopropyl-4H-1,2,4-triazol-3-yl)thio]acetic acid
Supplier: Matrix Scientific
077376-1G
077376-5G
077376-500MG
102665-898EA
484.62
USD
102665-898
102665-900
102665-896
[(5-Isopropyl-4H-1,2,4-triazol-3-yl)thio]acetic acid
[(5-Isopropyl-4H-1,2,4-triazol-3-yl)thio]acetic acid
Hazard: Irritant
Formula:
C₇H₁₁N₃O₂S |
MDL Number:
MFCD08457677 CAS Number: 905810-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...