3-(Methylthio)-5-(trifluoromethyl)-4H-1,2,4-triazol-4-amine

Supplier: Matrix Scientific

077375-5G 077375-500MG 077375-1G
102665-890EA 387.69 USD
102665-890 102665-892 102665-894
3-(Methylthio)-5-(trifluoromethyl)-4H-1,2,4-triazol-4-amine
3-(Methylthio)-5-(trifluoromethyl)-4H-1,2,4-triazol-4-amine

Hazard: Irritant

Formula: C₄H₅F₃N₄S
MDL Number: MFCD00277904
CAS Number: 164352-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR