1-(3,5-Difluorobenzyl)-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

077367-500MG 077367-5G 077367-1G
102665-844EA 484.62 USD
102665-844 102665-846 102665-842
1-(3,5-Difluorobenzyl)-5-oxopyrrolidine-3-carboxylic acid
1-(3,5-Difluorobenzyl)-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₁F₂NO₃
MDL Number: MFCD11655669
CAS Number: 1098340-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR