Ethyl-2,4-dichloro-5-(chlorosulfonyl)benzoate

Supplier: Matrix Scientific

077365-1G 077365-500MG 077365-5G
102665-832EA 323.08 USD
102665-832 102665-834 102665-830
Ethyl-2,4-dichloro-5-(chlorosulfonyl)benzoate
Ethyl-2,4-dichloro-5-(chlorosulfonyl)benzoate

Hazard: Irritant

Formula: C₉H₇Cl₃O₄S
MDL Number: MFCD11057851
CAS Number: 924859-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR