Ethyl-2-piperidin-1-ylpropanoate
Supplier: Matrix Scientific
077356-500MG
077356-1G
077356-5G
102665-780EA
743.08
USD
102665-780
102665-778
102665-776
Ethyl-2-piperidin-1-ylpropanoate
Ethyl-2-piperidin-1-ylpropanoate
Hazard: Irritant
Formula:
C₁₀H₁₉NO₂ |
MDL Number:
MFCD00956471 CAS Number: 63909-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...