Methyl-5-[(4-acetylphenoxy)methyl]-2-furoate
Supplier: Matrix Scientific
077353-500MG
077353-5G
077353-1G
102665-760EA
323.08
USD
102665-760
102665-758
102665-762
Methyl-5-[(4-acetylphenoxy)methyl]-2-furoate
Methyl-5-[(4-acetylphenoxy)methyl]-2-furoate
Hazard: Irritant
Formula:
C₁₅H₁₄O₅ |
MDL Number:
MFCD04967303 CAS Number: 832739-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...