(2Z)-3-(5-Chloro-1H-1,2,4-triazol-3-yl)acrylic acid
Supplier: Matrix Scientific
077338-5G
077338-500MG
077338-1G
102665-672EA
516.92
USD
102665-672
102665-674
102665-676
(2Z)-3-(5-Chloro-1H-1,2,4-triazol-3-yl)acrylic acid
(2Z)-3-(5-Chloro-1H-1,2,4-triazol-3-yl)acrylic acid
Hazard: Irritant
Formula:
C₅H₄ClN₃O₂ |
MDL Number:
MFCD20441344 CAS Number: 1322627-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...