2-(Isopropylamino)-4-methyl-1,3-thiazole-5-carboxylic acid
Supplier: Matrix Scientific
077337-500MG
077337-1G
102665-670EA
484.62
USD
102665-670
102665-668
2-(Isopropylamino)-4-methyl-1,3-thiazole-5-carboxylic acid
2-(Isopropylamino)-4-methyl-1,3-thiazole-5-carboxylic acid
Hazard: Irritant
Formula:
C₈H₁₂N₂O₂S |
MDL Number:
MFCD08165212 CAS Number: 927983-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...