7-[(4-Oxo-4,5-dihydro-1,3-thiazol-2-yl)amino]heptanoic acid
Supplier: Matrix Scientific
077329-500MG
077329-1G
102665-638EA
484.62
USD
102665-638
102665-636
7-[(4-Oxo-4,5-dihydro-1,3-thiazol-2-yl)amino]heptanoic acid
7-[(4-Oxo-4,5-dihydro-1,3-thiazol-2-yl)amino]heptanoic acid
Hazard: Irritant
Formula:
C₁₀H₁₆N₂O₃S |
MDL Number:
MFCD04141975 CAS Number: 797027-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...