2-Amino-5,5-bis(hydroxymethyl)-1,3-thiazolidin-4-one
Supplier: Matrix Scientific
077327-500MG
077327-5G
077327-1G
102665-628EA
646.15
USD
102665-628
102665-626
102665-630
2-Amino-5,5-bis(hydroxymethyl)-1,3-thiazolidin-4-one
2-Amino-5,5-bis(hydroxymethyl)-1,3-thiazolidin-4-one
Hazard: Irritant
Formula:
C₅H₁₀N₂O₃S |
MDL Number:
MFCD19982151 CAS Number: 1313822-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...