2-[(4-Methylpiperazin-1-yl)methyl]pyridin-3-ol
Supplier: Matrix Scientific
077324-1G
077324-500MG
077324-5G
102665-608EA
387.69
USD
102665-608
102665-612
102665-610
2-[(4-Methylpiperazin-1-yl)methyl]pyridin-3-ol
2-[(4-Methylpiperazin-1-yl)methyl]pyridin-3-ol
Hazard: Irritant
Formula:
C₁₁H₁₇N₃O |
MDL Number:
MFCD00641887 CAS Number: 95380-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...