Methyl-3-(1H-benzimidazol-1-yl)propanoate

Supplier: Matrix Scientific

077308-500MG 077308-1G
102665-530EA 387.69 USD
102665-530 102665-532
Methyl-3-(1H-benzimidazol-1-yl)propanoate
Methyl-3-(1H-benzimidazol-1-yl)propanoate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
MDL Number: MFCD02128778
CAS Number: 144186-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR