4-(5-Mercapto-4H-1,2,4-triazol-3-yl)phenol
Supplier: Matrix Scientific
077291-1G
077291-500MG
102665-442EA
387.69
USD
102665-442
102665-444
4-(5-Mercapto-4H-1,2,4-triazol-3-yl)phenol
4-(5-Mercapto-4H-1,2,4-triazol-3-yl)phenol
Hazard: Irritant
Formula:
C₈H₇N₃OS |
MDL Number:
MFCD01930057 CAS Number: 3414-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...