N-(2-Amino-5-methylphenyl)acetamide

Supplier: Matrix Scientific

077213-500MG
102665-062EA 387.69 USD
102665-062
N-(2-Amino-5-methylphenyl)acetamide
N-(2-Amino-5-methylphenyl)acetamide

Hazard: Irritant

Formula: C₉H₁₂N₂O
MDL Number: MFCD09221899
CAS Number: 43154-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR