2-[4-(Dimethylamino)phenyl]-7-(trifluoromethyl)-1,2,4-benzotriazin-3(2H)-one

Supplier: Matrix Scientific

077208-500MG
102665-056EA 193.85 USD
102665-056
2-[4-(Dimethylamino)phenyl]-7-(trifluoromethyl)-1,2,4-benzotriazin-3(2H)-one
2-[4-(Dimethylamino)phenyl]-7-(trifluoromethyl)-1,2,4-benzotriazin-3(2H)-one

Hazard: Irritant

Formula: C₁₆H₁₃F₃N₄O
MDL Number: MFCD20441328
CAS Number: 1338494-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR