4-(4-Bromobenzyl)tetrahydro-2H-pyran-4-carboxylic acid

Supplier: Matrix Scientific

077192-5G 077192-1G 077192-500MG
102664-970EA 646.15 USD
102664-970 102664-968 102664-972
4-(4-Bromobenzyl)tetrahydro-2H-pyran-4-carboxylic acid
4-(4-Bromobenzyl)tetrahydro-2H-pyran-4-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅BrO₃
MDL Number: MFCD17600247
CAS Number: 1338494-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR