4-(2-Fluorobenzyl)tetrahydro-2H-pyran-4-carboxylic acid
Supplier: Matrix Scientific
077188-1G
077188-5G
077188-500MG
102664-948EA
1938.46
USD
102664-948
102664-946
102664-944
4-(2-Fluorobenzyl)tetrahydro-2H-pyran-4-carboxylic acid
4-(2-Fluorobenzyl)tetrahydro-2H-pyran-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₅FO₃ |
MDL Number:
MFCD17404694 CAS Number: 1272802-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...