2-{[(3-Chloro-2-methylphenyl)amino]methyl}-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindole-1,3-dione

Supplier: Matrix Scientific

077180-1G 077180-500MG
102664-904EA 323.08 USD
102664-904 102664-902
2-{[(3-Chloro-2-methylphenyl)amino]methyl}-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindole-1,3-dione
2-{[(3-Chloro-2-methylphenyl)amino]methyl}-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindole-1,3-dione

Hazard: Irritant

Formula: C₁₇H₁₇ClN₂O₂
MDL Number: MFCD17069389
CAS Number: 1241675-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR