4-(1,3-Dioxooctahydro-4,6-ethenocyclopropa[f]isoindol-2(1H)-yl)benzoic acid

Supplier: Matrix Scientific

077152-500MG 077152-1G 077152-5G
102664-748EA 743.08 USD
102664-748 102664-746 102664-744
4-(1,3-Dioxooctahydro-4,6-ethenocyclopropa[f]isoindol-2(1H)-yl)benzoic acid
4-(1,3-Dioxooctahydro-4,6-ethenocyclopropa[f]isoindol-2(1H)-yl)benzoic acid

Hazard: Irritant

Formula: C₁₈H₁₅NO₄
MDL Number: MFCD01142190
CAS Number: 354812-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR