(4,5,6,7,8,8-Hexachloro-1,3-dioxo-1,3,3a,4,7,7a-hexahydro-2H-4,7-methanoisoindol-2-yl)acetic acid

Supplier: Matrix Scientific

077150-1G 077150-500MG 077150-5G
102664-736EA 743.08 USD
102664-736 102664-734 102664-732
(4,5,6,7,8,8-Hexachloro-1,3-dioxo-1,3,3a,4,7,7a-hexahydro-2H-4,7-methanoisoindol-2-yl)acetic acid
(4,5,6,7,8,8-Hexachloro-1,3-dioxo-1,3,3a,4,7,7a-hexahydro-2H-4,7-methanoisoindol-2-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₅Cl₆NO₄
MDL Number: MFCD02763354
CAS Number: 60852-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR