4-Methyl-2-(4,5,6,7-tetrachloro-1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid
Supplier: Matrix Scientific
077147-5G
077147-500MG
077147-1G
102664-718EA
613.85
USD
102664-718
102664-716
102664-714
4-Methyl-2-(4,5,6,7-tetrachloro-1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid
4-Methyl-2-(4,5,6,7-tetrachloro-1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid
Hazard: Irritant
Formula:
C₁₄H₁₁Cl₄NO₄ |
MDL Number:
MFCD02319303 CAS Number: 90027-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...