6,7-Dimethoxy-2-methyl-3-[4-(methylthio)phenyl]-1-oxo-1,2,3,4-tetrahydroisoquinoline-4-carboxylic acid

Supplier: Matrix Scientific

077079-500MG 077079-1G
102664-388EA 516.92 USD
102664-388 102664-390
6,7-Dimethoxy-2-methyl-3-[4-(methylthio)phenyl]-1-oxo-1,2,3,4-tetrahydroisoquinoline-4-carboxylic acid
6,7-Dimethoxy-2-methyl-3-[4-(methylthio)phenyl]-1-oxo-1,2,3,4-tetrahydroisoquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₂₁NO₅S
MDL Number: MFCD12027136
CAS Number: 896620-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR