2-Bromo-4,5-difluorobenzotrifluoride ≥95%
Supplier: Matrix Scientific
077069-1G
102664-366EA
320
USD
102664-366
2-Bromo-4,5-difluorobenzotrifluoride ≥95%
2-Bromo-4,5-difluorobenzotrifluoride
Hazard: Irritant
Formula:
C₇H₂BrF₅ |
MDL Number:
MFCD11226699 CAS Number: 112290-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...