tert-Butyl-3-[methoxy(methyl)carbamoyl]cyclohexane-1-carboxylate ≥97%
Supplier: Matrix Scientific
077053-500MG
077053-1G
102664-336EA
504.62
USD
102664-336
102664-334
tert-Butyl-3-[methoxy(methyl)carbamoyl]cyclohexane-1-carboxylate ≥97%
tert-Butyl-3-[methoxy(methyl)carbamoyl]cyclohexane-1-carboxylate
Hazard: Irritant
Formula:
C₁₄H₂₅NO₄ MW: 271.36 g/mol |
MDL Number:
MFCD22383958 CAS Number: 1393845-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...