4-{[4-(Hydroxymethyl)pyrimidin-2-yl]amino}benzonitrile ≥97%
Supplier: Matrix Scientific
077033-1G
077033-500MG
102664-262EA
1504.62
USD
102664-262
102664-260
4-{[4-(Hydroxymethyl)pyrimidin-2-yl]amino}benzonitrile ≥97%
4-{[4-(Hydroxymethyl)pyrimidin-2-yl]amino}benzonitrile
Hazard: Irritant
Formula:
C₁₂H₁₀N₄O Melting Pt: 172…174 °C |
MDL Number:
MFCD22383942 CAS Number: 1393845-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...