Ethyl-7-methyl-7H-pyrrolo[2,3-d]pyrimidine-4-carboxylate ≥97%

Supplier: Matrix Scientific

077032-500MG 077032-1G
102664-256EA 600 USD
102664-256 102664-258
Ethyl-7-methyl-7H-pyrrolo[2,3-d]pyrimidine-4-carboxylate ≥97%
Ethyl-7-methyl-7H-pyrrolo[2,3-d]pyrimidine-4-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₂
MW: 205.22 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD22383951
CAS Number: 1393845-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR