N,N-Dimethyl-2-(2-oxocyclopentyl)acetamide ≥97%

Supplier: Matrix Scientific

077024-1G 077024-500MG
102664-226EA 504.62 USD
102664-226 102664-224
N,N-Dimethyl-2-(2-oxocyclopentyl)acetamide ≥97%
N,N-Dimethyl-2-(2-oxocyclopentyl)acetamide

Hazard: Irritant

Formula: C₉H₁₅NO₂
MDL Number: MFCD22383948
CAS Number: 1393845-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR