2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-1,3-benzodiazole ≥95%
Supplier: Matrix Scientific
077013-500MG
077013-1G
102664-182EA
504.62
USD
102664-182
102664-180
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-1,3-benzodiazole ≥95%
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-1,3-benzodiazole
Hazard: Irritant
Formula:
C₁₃H₇ClF₃N₃ |
MDL Number:
MFCD22383952 CAS Number: 1393845-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...