5-Bromo-4-methoxy-7-methyl-7H-pyrrolo[2,3-d]pyrimidine ≥95%

Supplier: Matrix Scientific

077007-1G 077007-500MG
102664-160EA 749.23 USD
102664-160 102664-158
5-Bromo-4-methoxy-7-methyl-7H-pyrrolo[2,3-d]pyrimidine ≥95%
5-Bromo-4-methoxy-7-methyl-7H-pyrrolo[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₈H₈BrN₃O
Melting Pt: 151…153 °C
MDL Number: MFCD22383939
CAS Number: 1393845-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR