5-Bromo-3-N-cyclopentylpyridine-2,3-diamine ≥97%

Supplier: Matrix Scientific

077005-500MG 077005-1G
102664-152EA 504.62 USD
102664-152 102664-150
5-Bromo-3-N-cyclopentylpyridine-2,3-diamine ≥97%
5-Bromo-3-N-cyclopentylpyridine-2,3-diamine

Hazard: Irritant

Formula: C₁₀H₁₄BrN₃
Melting Pt: 56…58 °C
MDL Number: MFCD22383937
CAS Number: 1393845-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR