1H-Pyrrolo[2,3-c]pyridin-7-ylmethanol ≥97%

Supplier: Matrix Scientific

Synonyms: {1H-pyrrolo[2,3-c]pyridin-7-yl}methanol

076661-500MG
102547-322EA 1396.92 USD
102547-322
1H-Pyrrolo[2,3-c]pyridin-7-ylmethanol ≥97%
1H-Pyrrolo[2,3-c]pyridin-7-ylmethanol

Hazard: Irritant

Formula: C₈H₈N₂O
MW: 148.17 g/mol
Melting Pt: 180 °C
MDL Number: MFCD22373669
CAS Number: 1379526-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR