N-Methyl-1H,4H,5H,6H-cyclopenta[c]pyrazole-3-carboxamide ≥97%
Supplier: Matrix Scientific
076644-1G
076644-500MG
102547-250EA
803.08
USD
102547-250
102547-252
N-Methyl-1H,4H,5H,6H-cyclopenta[c]pyrazole-3-carboxamide ≥97%
N-Methyl-1H,4H,5H,6H-cyclopenta[c]pyrazole-3-carboxamide
Hazard: Irritant
Formula:
C₈H₁₁N₃O MW: 165.2 g/mol Melting Pt: >250 °C |
MDL Number:
MFCD22373665 CAS Number: 1335131-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...