4-Hydrazinyl-2,6-bis(trifluoromethyl)pyridine ≥95%
Supplier: Matrix Scientific
076638-500MG
102547-222EA
1396.92
USD
102547-222
4-Hydrazinyl-2,6-bis(trifluoromethyl)pyridine ≥95%
4-Hydrazinyl-2,6-bis(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₇H₅F₆N₃ Melting Pt: 124…126 °C |
MDL Number:
MFCD01936019 CAS Number: 240115-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...